Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC JH295 25mg | 1311143-71-1 | 320.35 g/mol | C18H16N4O2 | 25 MG
SDP

Supplier:  Medchemexpress LLC HY11642325MG

Encompass_Preferred

This compound (CAS 1311143-71-1) is a small-molecule, cysteine-targeted inhibitor of NIMA-related kinase 2 (Nek2) that irreversibly inactivates the enzyme via alkylation of a catalytic cysteine. It is supplied as a solid for research use; solutions should be prepared fresh due to limited stability in solution.

  • Potent, irreversible Nek2 inhibition with reported IC50 ≈ 770 nM.
  • Molecular formula C18H16N4O2; molecular weight 320.35 g/mol.
  • High purity (98.66%) and yellow to orange solid appearance.
  • Soluble in DMSO (~55 mg/mL); sonication and newly opened DMSO recommended.
  • Store at -20°C under inert atmosphere; avoid extended storage in solution.

Catalog No. 50-002-27216


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.