Learn More
Medchemexpress LLC JH295 hydrate 1mg | 2714087-26-8 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY116423A1MG
Encompass_Preferred
JH295 hydrate is a potent, irreversible, and selective NIMA-related kinase 2 (Nek2) inhibitor that alkylates Cys22 and inhibits Nek2 with an IC50 of 770 nM. It contains an alkyne functional group for copper-catalyzed azide-alkyne cycloaddition (click chemistry) and is supplied as a solid with ≥99.0% purity for biochemical and cellular studies.
  • Irreversible Nek2 inhibitor (IC50 770 nM).
  • Alkylates Cys22 for covalent target engagement.
  • Contains an alkyne handle for click-chemistry applications.
  • High purity (≥99.0%) suitable for research use.
  • Stable as a powder under recommended storage conditions.
Catalog No. 50-002-28168