Learn More
ACCEGEN BIOTECH JHH-7
Supplier:  ACCEGEN BIOTECH ABCTC0475
Encompass
HBV integrated human hepatocellular carcinoma cell line. HBV integrated human HBs-Ag positive hepatocellular carcinoma (Ed.III) with liver cirrhosis. Procuces albumin, AFP, CEA and ferritin.
Catalog No. NC1984858