Learn More
Creative Bioarray JHOM-1 Cells, Frozen, 1x10^6 cells/1 mL, Adherent, For Research Use Only
Supplier: Creative Bioarray CSCC6615J
The Creative Bioarray JHOM-1 Cells are a cryopreserved human ovarian epithelial tumor cell line derived from a female patient with mucinous ovarian cancer. Designed specifically for cancer research applications, this adherent cell line provides a valuable in vitro model for studying the mechanisms of ovarian cancer, evaluating drug efficacy, and aiding therapeutic development.
- Derived from a female donor with ovarian epithelial tumor
- Species: Human
- Growth properties: Adherent
- Application: For research use only
- Quantity: 1 x 106 cells / 1.0 ml
- Format: Frozen
- Doubling time: 60 hours
- Available genomic data: Whole exome sequencing, whole genome sequencing
- Available proteomic data: Reverse-phase protein array
- Available transcriptomic data: Microarray, RNAseq
- Included in Cancer Dependency Map and MD Anderson Cell Lines Project
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.