Learn More
Medchemexpress LLC HY-122218 10mg Medchemexpress, JHU-083 CAS:1998725-11-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12221810MG
Encompass_Preferred
Medchemexpress, HY-122218 10mg JHU-083 CAS:1998725-11-3 JHU-083, a prodrug of 6-diazo-5-oxo-L-norleucine (DON; HY-108357), is an orally active and selective glutaminase antagonist. JHU-083 blocks glutaminase activity in brain CD11b+ cells and experimental cerebral malaria (ECM) resulting in a net decrease of glutamate levels in the animals[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8865