Learn More
Selleck Chemical LLC JIB-04
SDP
Supplier:  Selleck Chemical LLC S72811G
Encompass_Preferred
JIB-04 (NSC 693627) is a pan-selective Jumonji histone demethylase inhibitor with IC50 of 230 340 855 445 435 1100 and 290 nM for JARID1A JMJD2E JMJD3 JMJD2A JMJD2B JMJD2C and JMJD2D in cell-free assays respectively JIB 04 also induces cell apoptosis
Catalog No. 50-000-04538