Learn More
Medchemexpress LLC HY-13307 5mg Medchemexpress, JK184 CAS:315703-52-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY133075MG
Encompass_Preferred
Medchemexpress, HY-13307 5mg JK184 CAS:315703-52-7 JK184 is a potent Hedgehog (Hh) pathway inhibitor with IC50 of 30 nM in mammalian cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1328