Learn More
Medchemexpress LLC HY-138153 5mg , JKE-1674 CAS:2421119-60-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1381535MG
Encompass_Preferred
HY-138153 5mg JKE-1674 CAS: 2421119-60-8 JKE-1674 is an orally active glutathione peroxidase 4 (GPX4) inhibitor and an active metabolite of GPX4 inhibitor ML-210. JKE-1674, an analog of ML-210 in which the nitroisoxazole ring is replaced with an α-nitroketoxime. JKE-1674 can convert into a nitrile oxide JKE-1777. JKE-1674 kills LOX-IMVI cells in a manner that is equipotent to ML-210 and is completely rescued by ferroptosis inhibitors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9353