Learn More
INTACT GENOMICS INC JM109 (DE3) Chemically Competent Cells 12x50µl
Supplier:  INTACT GENOMICS INC 106312
Encompass
Intact Genomics JM109-DE3 chemically competent E. coli cells are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning. JM109-DE3 is useful for the high-level expression of genes cloned into vectors for the expression of sequence downstream from the T7 promoter.ig™ JM109-DE3 chemically competent cells have the following features: • recA– and endA– to minimize recombination and improve the quality of plasmid DNA. • Contains F’ episome for blue/white screening.Product Includes & Storage:1) ig™ JM109(DE3) competent cells: -80 ºC2) pUC19 control DNA: -20 ºC3) Recovery medium: 4 ºC
Catalog No. 50-000-82883