Learn More
TARGETMOL CHEMICALS INC JMJD6 INHIBITOR WL12 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T99395MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JMJD6 inhibitor WL12 (ZINC6733033) is a first-in-class JMJD6 inhibitor and was shown to be able to suppress JMJD6-dependent cancer cell proliferation including cervical and liver cancer cells providing a small-molecule propurity: 98%
Catalog No. 50-257-8924