Learn More
Medchemexpress LLC JNJ-10397049 | 708275-58-5 | 99.2% | 484.18 | 5 MG

Supplier: Medchemexpress LLC 5MGHY10896
JNJ-10397049 is a potent and selective orexin 2 receptor (OX2R) antagonist, with a pKi of 8.3. It is 600-fold selective for the OX2R over the OX1R. For research use only. In vivo, JNJ-10397049 (10-30 mg/kg) decreases the latency for persistent sleep and increased nonrapid eye movement and rapid eye movement sleep time. It also blocks ethanol self-administration, place preference, and reinstatement.
- Decreases the latency for persistent sleep and increases nonrapid eye movement and rapid eye movement sleep time.
- Blocks ethanol self-administration, place preference, and reinstatement.
- In vitro solubility: DMSO: 170 mg/mL (351.11 mM).
- In vivo suspended solution protocol: 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline; solubility: 4.25 mg/mL (8.78 mM).
- In vivo clear solution protocol: 10% DMSO, 90% Corn Oil; solubility: ≥ 4.25 mg/mL (8.78 mM).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.