Learn More
Medchemexpress LLC JNJ-26070109 | 844645-08-5 | 99.3% | 547.37 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11131310MM/1ML
Encompass_Preferred
JNJ-26070109 is a high-affinity, competitive, orally bioactive, and selective cholecystokinin 2 (CCK2) receptor antagonist with good pharmacokinetic properties. It targets CCK2 receptors that regulate gastric acid secretion and growth of the gastrointestinal mucosa, making it a promising tool for gastroesophageal reflux disease research.
  • High-affinity, competitive, orally bioactive, and selective.
  • Good pharmacokinetic properties.
  • Targets CCK2 receptors that regulate gastric acid secretion.
  • Supports research into gastroesophageal reflux disease.
Catalog No. 50-003-43782