Learn More
Medchemexpress LLC JNJ-28583113 | 2765255-93-2 | >98.8% | 366.38 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY149143
Encompass_Preferred
JNJ-28583113 is a brain-permeable TRPM2 antagonist. It inhibits TRPM2-blocked phosphorylation of GSK3α and β subunits, protects cells from oxidative stress-induced cell death, and suppresses cytokine release in response to pro-inflammatory stimuli in microglia.
  • Inhibits TRPM2 in cells (IC50=100 nM chimpanzee, 25 nM rat, 126 nM human)
  • Prevents cells from H2O2 induced cell death up to 1 mM of H2O2
  • Protects HeLa cells from H2O2 induced morphological changes
  • Brain penetrant and achieves 400 ng/mL in the brain compartment
Catalog No. 50-003-63341