Learn More
Medchemexpress LLC JNJ-38877605 | 943540-75-8 | 100.0 % | C19H13F2N7 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY50683
Encompass_Preferred
JNJ-38877605 is an orally active ATP-competitive inhibitor of c-Met with an IC50 of 4 nM, demonstrating 600-fold selectivity for c-Met over 200 other tyrosine and serine-threonine kinases. It inhibits c-Met phosphorylation and regulates lipid accumulation, making it suitable for research in tumor and metabolic diseases.
  • Orally active ATP-competitive c-Met inhibitor
  • IC50 of 4 nM for c-Met
  • 600-fold selectivity for c-Met over 200 other kinases
  • Inhibits c-Met phosphorylation
  • Regulates lipid accumulation
  • Suitable for tumor and metabolic disease research
Catalog No. 50-003-58895