Learn More
Selleck Chemical LLC JNJ-38877618 S8854-5MG
SDP
Supplier:  Selleck Chemical LLC S88545MG
Encompass_Preferred
JNJ-38877618 (OMO-1) is a potent highly selective orally bioavailable Met (c-Met) kinase inhibitor with binding affinity (Kd) of 1 4 nM and enzyme inhibitory activity against wt and M1268T mutant Met (c-Met) (2 and 3 nM IC50)
Catalog No. 50-000-25156