Learn More
Medchemexpress LLC JNJ-42041935 | 1193383-09-3 | 99.8% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1283210MG
Encompass_Preferred
JNJ-42041935 is a potent, competitive, and selective inhibitor of prolyl hydroxylase (PHD) enzymes, including PHD1, PHD2, and PHD3. It has shown effectiveness in reversing inflammation-induced anemia in rats and increasing reticulocytes, hemoglobin, and hematocrit in in vivo studies.
  • Potent, competitive, and selective inhibitor of prolyl hydroxylase
  • Inhibits PHD1, PHD2, and PHD3
  • Most potent inhibitor of PHD2181-417
  • Effectively reverses inflammation-induced anemia in rats
  • Increases reticulocytes, hemoglobin, and hematocrit in vivo
Catalog No. 50-003-39209