Learn More
Medchemexpress LLC JNJ-42041935 | 1193383-09-3 | 99.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY12832100MG
Encompass_Preferred
JNJ-42041935 is described as a potent, competitive, and selective inhibitor of prolyl hydroxylase (PHD), demonstrating inhibition of PHD1, PHD2, and PHD3 with specific pKi values. It has been investigated for its efficacy in reversing inflammation-induced anemia in rat models and for increasing reticulocytes, hemoglobin, and hematocrit in vivo.
  • Potent inhibition of specific enzymes.
  • Competitive and selective mode of action.
  • Effective in reversing anemia in preclinical models.
  • Increases red blood cell precursors, hemoglobin, and hematocrit in animal studies.
Catalog No. 50-003-42333