Learn More
Medchemexpress LLC HY-19559 5mg Medchemexpress, JNJ-46778212 CAS:1363281-27-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY195595MG
Encompass_Preferred
Medchemexpress, HY-19559 5mg JNJ-46778212 CAS:1363281-27-9 JNJ-46778212 (VU 0409551) is an mGlu5 positive allosteric modulator with an EC50 of 260 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2145