Learn More
Medchemexpress LLC JNJ-47117096 hydrochloride | 1610536-69-0 | 99.7% | 398.89 g·mol⁻1 | C21H23ClN4O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY1242010MG
Encompass_Preferred
JNJ-47117096 hydrochloride is the hydrochloride salt of a potent and selective MELK inhibitor, with reported MELK IC50 = 23 nM and Flt3 IC50 = 18 nM. It is supplied as a high-purity research reagent for in vitro kinase profiling and cancer-signaling studies; not for human use.
  • Potent MELK inhibition (IC50 = 23 nM).
  • Secondary Flt3 activity (IC50 = 18 nM).
  • Hydrochloride salt form for improved solubility and stability.
  • High reported purity (≈99.7%).
  • Suitable for in vitro kinase assays and cell signaling experiments.
  • Intended for research use only.
Catalog No. 50-002-36780