Learn More
Medchemexpress LLC JNJ-47117096 hydrochloride | 1610536-69-0 | 99.7% | 398.89 g/mol | C21H23ClN4O2 | 100MG
SDP
Supplier:  Medchemexpress LLC HY12420100MG
Encompass_Preferred
JNJ-47117096 hydrochloride is a research-grade small-molecule kinase inhibitor supplied as a hydrochloride salt. It potently inhibits maternal embryonic leucine zipper kinase (MELK) and also inhibits Flt3, making it suitable for biochemical and cellular studies of kinase signaling pathways.
  • Potent MELK inhibitor (IC50 23 nM).
  • Also inhibits Flt3 (IC50 18 nM).
  • High purity suitable for research (99.7%).
  • Solid white to off-white appearance for easy handling.
  • Molecular weight 398.89 g/mol and formula C21H23ClN4O2.
  • Available in multiple package sizes from 5 mg to 500 mg.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-37798