Learn More
Cayman Chemical JNJ-64619178 1mg
SDP
Supplier:  Cayman Chemical 292311 MG
Encompass_Preferred
A PRMT5 inhibitor (IC50 = 0.14 nM); inhibits the growth of various cancer cells in vitro and reduces tumor growth in NSCLC and SCLC mouse xenograft models
Catalog No. 50-293-7162