Learn More
Selleck Chemical LLC JNJ-A07-E1422-100MG
SDP
Supplier:  Selleck Chemical LLC E1422100MG
Encompass_Preferred
JNJ-A07 is a highly potent inhibitor of Dengue virus (DENV) with an EC50 of 0 1 nM in the DENV-2 strain (16681) in Vero cells It exhibits antiviral activity by preventing the formation of the viral replication complex through blocking the interaction between two viral proteins NS3 and NS4B
Catalog No. 50-313-4939