Learn More
Medchemexpress LLC Jnj DGAT2-A | 1962931-71-0 | MFCD30182366 | 98.3% | 523.38 g·mol⁻1 | C24H16BrFN4O2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1103815MG
Encompass_Preferred
JNJ-DGAT2-A is a selective diacylglycerol acyltransferase 2 (DGAT2) inhibitor used for research into triglyceride synthesis and lipid metabolism. It exhibits potent biochemical activity and cellular efficacy, and is supplied at analytical purity suitable for in vitro studies.
  • Selective DGAT2 inhibition with an IC50 of 0.14 μM.
  • Reduces triglyceride species in HepG2 cells with IC50 values ~0.66-0.99 μM.
  • High purity (≈98.3%) appropriate for biochemical assays.
  • Available in research-scale sizes, including 5 mg.
  • Stable when stored dry at 4°C; recommended low-temperature storage for solutions.
Catalog No. 50-002-63632