Learn More
Medchemexpress LLC JNK-IN-13 | 345986-38-1 | 97.6% | C13H7ClN4S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY77009
Encompass_Preferred
JNK-IN-13 is a potent and selective JNK inhibitor that exhibits IC50s of 290 nM for JNK3 and 500 nM for JNK2. This product is intended for research use only and appears as a solid, typically white to yellow in color.
  • Potent and selective JNK inhibitor
  • IC50s of 290 nM for JNK3 and 500 nM for JNK2
  • For research use only
  • Appears as a solid
  • White to yellow color
  • Store at 4°C under nitrogen
  • If in solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen
Catalog No. 50-003-57870