Learn More
Apexbio Technology LLC JNK-IN-7, 50mg. Cas: 1408064-71-0 MFCD: MFCD25976765. Selective JNK inhibitor
SDP
Supplier:  Apexbio Technology LLC A351950
Encompass_Preferred
JNK-IN-7 is a selective JNK inhibitor with IC50 values of 1.54 nM, 1.99 nM, 0.75 nM to JNK1, JNK2, JNK3, respectively. It also inhibits phosphorylation of c-Jun, which is a direct substrate of JNK kinase. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4809