Learn More
Selleck Chemical LLC JNK Inhibitor IX 10mg 312917-14-9 TCS JNK 5a
SDP
Supplier:  Selleck Chemical LLC S750810MG
Encompass_Preferred
JNK inhibitor IX (TCS JNK 5a) is a selective and potent JNK inhibitor with pIC50 of 6.5 and 6.7 for JNK2 and JNK3, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4874