Learn More
Selleck Chemical LLC JNK Inhibitor VIII
SDP
Supplier:  Selleck Chemical LLC S77945MG
Encompass_Preferred
JNK Inhibitor VIII (TCS JNK 6o) is an inhibitor of c-Jun N-terminal kinases with IC50 of 45 nM and 160 nM for JNK-1 and JNK-2 respectively JNK Inhibitor VIII (TCS JNK 6o) inihibits JNK-1 JNK-2 and JNK-3 with Ki of 2 nM 4 nM and 52 nM respectively
Catalog No. 50-000-01072