Learn More
Cayman Chemical JNK InhIbItr IX 50mg
SDP
Supplier:  Cayman Chemical 1562450 MG
Encompass_Preferred
A thienylnaphthamide compound that selectively targets the ATP binding site of JNK2 and JNK3 (pIC50s = 6.5 and 6.7, respectively)
Catalog No. 50-297-1483