Learn More
Cayman Chemical JNK InhIbItr VII 1mg
SDP
Supplier:  Cayman Chemical 288311 MG
Encompass_Preferred
A non-selective JNK inhibitor (IC50s = 1.54, 1.99, and 0.75 nM for JNK1, -2, and -3, respectively); inhibits phosphorylation of c-Jun in HeLa and A375 cells (EC50s = 130 and 244 nM, respectively)
Catalog No. 50-294-0049