Learn More
Chemglass Life Sciences Joint, 28/12, Polished Socket, Clamp Size 28, 16mm Tubing
SDP
Supplier:  Chemglass Life Sciences CG121P03
Encompass_Preferred
Joint, 28/12, Polished Socket, Clamp Size 28, 16mm Tubing. For use with CG-122, o-ring ball joint to provide for a flexible high vacuum seal. Polished sealing surface does not abrade o-ring. Note: Not for use with CG-120 ground ball joints.
Catalog No. 50-153-1059