Learn More
Chemglass Life Sciences 28/15 Ball Joint, Clamp Size: 28, Tubing OD: 18.80-19.20mm
SDP
Supplier:  Chemglass Life Sciences CG12008
Encompass_Preferred
28/15 Ball Joint, Clamp Size: 28, Tubing OD: 18.80-19.20mm. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.
Catalog No. 50-153-1053