Learn More
Med Vet International Jorvet 20g x 8in. Spinal Needle, J0529M
Supplier: Med Vet International JOR529M
Jorvet 20g x 8in. spinal needle, is used as in gerneral purpose aspiration biopsies and drainage procedures. Each needle has a matched ground bevel that eliminates the possibility of tissue coring. They attach to a standard luer fitting and is sterile and not autoclavable.