Learn More
Med Vet International Jorvet Equine Leg Wrap, Individual, sterile, 12" x 36", J0849AS
Supplier:  Med Vet International JOR849AS
Encompass_Preferred
Made of soft quality bulk cotton which is covered on both sides with a permeable thin layer of nonwoven material. The wrap acts much like a diaper in that the wound exudate is lifted and absorbed into the thick middle cotton layer. A dark mark mid-line runs full length to give proper orientation during application. A wide variety of materials and sizes are now available:
Catalog No. 50-005-82180