Learn More
Med Vet International Jorvet Hypodermic Needle, 16g x 3in., J0174CG, 12/box
Supplier:  Med Vet International JOR174CG
Encompass_Preferred
Jorvet 16g x 3in. Hypodermic needle is made of stainless steel, has a round hub, and is packaged in a cardboard box or clear plastic case. This product contains 12 needles.
Catalog No. 50-005-82631