Learn More
Med Vet International Jorvet Hypodermic Needle, 20g x 2in., J0174AE, 12/box
Supplier:  Med Vet International JOR174AE
Encompass_Preferred
Jorvet 20g x 2in. Hypodermic needle is made of stainless steel, has a round hub, and is packaged in a cardboard box or clear plastic case. This product contains 12 needles.
Catalog No. 50-005-82531