Learn More
Jorgensen Labs JorVet Soda Lime, 5 gallon
SDP
Supplier:  Jorgensen Labs J0553B
Encompass_Preferred
JorVet soda lime is a high-grade granular product that effectively absorbs carbon dioxide that is produced as an anesthesia by-product. It is a mixture of calcium hydroxide Ca(OH)2 also known as lime plus a smaller amount of sodium hydroxide NaOH.
Catalog No. NC1348911