Learn More
Cayman Chemical JP104 Fatty Acids and Derivati
SDP
Supplier:  Cayman Chemical 1000866150 MG
Encompass_Preferred
An irreversible FAAH inhibitor of the carbamate class with an IC50 value of 7.3 nM for the human recombinant enzyme when tested using radiolabeled oleamide as the substrate; the alkyl derivative on JP104 reacts with azide-modified reporter tags, such as azido-rhodamine or azido-biotin, for visualization of JP104 bound to FAAH in vivo
Catalog No. 50-288-9009