Learn More
Medchemexpress LLC Jp1302 dihydrochloride | 1259314-65-2 | MFCD10565599 | 99.9% | 441.40 | C24H26Cl2N4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10321350MG
Encompass_Preferred
JP1302 dihydrochloride is a selective α2C-adrenoceptor antagonist used as a research reagent for receptor binding and pharmacological studies. The compound is supplied as the dihydrochloride salt and has reported high purity and sub-micromolar affinity for human α2C receptors.
  • Selective antagonist of α2C-adrenoceptor with high affinity (Ki = 28 ± 2 nM).
  • Markedly lower affinity at α2A and α2B subtypes, indicating subtype selectivity.
  • Reported high purity (99.94%).
  • Available in multiple pack sizes, including 50 mg.
  • Suitable for in vitro receptor binding and pharmacological assays.
Catalog No. 50-002-47208