Learn More
Selleck Chemical LLC JPH203 5mg 1037592-40-7 KYT-0353
SDP
Supplier:  Selleck Chemical LLC S86675MG
Encompass_Preferred
JPH203 (KYT-0353), a selective L-type amino acid transporter 1 inhibitor, shows a dramatic inhibition of leucine uptake (IC50=0.06 ¿M) and cell growth (IC50=4.1 ¿M) in human colon cancer cells (HT-29), human oral cancer cells (YD-38) and leukemic cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8065