Learn More
TARGETMOL CHEMICALS INC JPH203 dihydrochloride 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T117271MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. JPH203 dihydrochloride (KYT-0353 dihydrochloride) is a tyrosine analog, acts as a selective inhibitor of L-type amino acid transporter 1 (LAT1), and is used in cancer research. Purity 95.39%
Catalog No. 50-288-0611