Learn More
TARGETMOL CHEMICALS INC JQ-1 CARBOXYLIC ACID 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T544325MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JQ-1 (carboxylic acid) is a cell-permeable BRD4 inhibitor with IC50s of 77 nM for BRD4(1) and 33 nM for BRD4(2)purity: 99%
Catalog No. 50-257-6681