Learn More
Selleck Chemical LLC (+)-JQ1 10mg 1268524-70-4
SDP
Supplier:  Selleck Chemical LLC S711010MG
Encompass_Preferred
(+)-JQ1 is a BET bromodomain inhibitor, with IC50 of 77 nM/33 nM for BRD4(1/2) in cell-free assays, binding to all bromodomains of the BET family, but not to bromodomains outside the BET family. (+)-JQ1 suppresses cell proliferation via inducing autophagy. (+)-JQ1 inhibits the expression of Nuclear receptor binding SET domain protein 3 (NSD3) target genes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0713027