Learn More
Medchemexpress LLC (+)-JQ1 PA | 2115701-93-2 | 98.2% | 437.95 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY112789100MG
Encompass_Preferred
(+)-JQ1 PA is a derivative of the Bromodomain and extra-terminal (BET) inhibitor JQ1, with an IC50 of 10.4 nM. It functions as a click chemistry reagent, containing an Alkyne group, and is capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain Azide groups.
  • Functions as a derivative of a BET inhibitor.
  • Exhibits an IC50 of 10.4 nM.
  • Acts as a click chemistry reagent.
  • Contains an alkyne functional group.
  • Capable of copper-catalyzed azide-alkyne cycloaddition.
  • Reacts with molecules containing azide groups.
Catalog No. 50-004-37383