Learn More
Medchemexpress LLC Jqad1 | 2417097-18-6 | 98.2% | 932.95 | C48H52F4N6O9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1457655MG
Encompass_Preferred
JQAD1 is a CRBN-dependent proteolysis-targeting chimera (PROTAC) that selectively targets the histone acetyltransferase EP300 for degradation. It suppresses EP300 expression, reduces H3K27ac modification, and can induce apoptosis in cellular models, making it suitable as a research reagent for studying EP300 function and target validation.
  • Selective EP300 degradation in cell-based assays.
  • CRBN-dependent PROTAC mechanism of action.
  • Reduces H3K27ac histone modification associated with EP300 activity.
  • Reported high purity suitable for biological research.
  • Available in small quantities for experimental screening and validation.
Catalog No. 50-002-21133