Learn More
Apexbio Technology LLC JSH-23, 25mg. Cas: 749886-87-1 MFCD: MFCD09753595. NF-κB inhibitor
SDP
Supplier:  Apexbio Technology LLC B164525
Encompass_Preferred
JSH-23 is an inhibitor of NF-B transcriptional activity with IC50 value of 7.1M [1].JSH-23 is developed to inhibit NF-B transcriptional activity in LPS-stimulated macrophages RAW 264.7. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5277