Learn More
YMC America, Inc JsphrH80 4um 100x.15mm 1/16
SDP
Supplier:  YMC America, Inc JH08S0410F5AU
Encompass
The J sphere series of columns consists of three kinds of ODS made from the same silica gel base each with a different ligand coverage The 4 um 80 silica support provides outstanding chromatographic efficiency while the high surface area material (500 m2/g) offers exceptional interaction with analytes This combination provides excellent peak symmetry across all J sphere products The variations in ligand coverage produce three columns with different hydrophobicity hydrogen bonding capacity and steric selectivity for tertiary structures Because of this J sphere can be used with confidence to identify structural differences of solutes and to improve the efficiency of separation optimization
Catalog No. 50-008-80900