Learn More
Selleck Chemical LLC JTC-801 S2722-5mg
SDP
Supplier:  Selleck Chemical LLC S27225MG
Encompass_Preferred
JTC-801 is a selective opioid receptor-like1 (ORL1) receptor antagonist with IC50 of 94 nM weakly inhibits receptors and
Catalog No. 50-217-3097