Learn More
Cayman Chemical JTE-013, 383150-41-2, 1 mg
SDP
Supplier:  Cayman Chemical 100094581
Encompass_Preferred
JTE-013 is a potent, selective S1P2 receptor antagonist that binds to the human and rat receptors with IC50 values of 17 and 22 nM, respectively, (IC50 values >10 µM for human S1P1 and S1P3).{14560} It reverses the inhibitory effects of S1P on cell migration of vascular endothelial cells and smooth muscle cells. Similarly, JTE-013 reverses the inhibition of S1P on invasion and migration of B16 melanoma cells.{14560,14559} JTE-013 inhibits S1P-induced contraction of, as well as cAMP accumulation in, coronary artery smooth muscle cells.{14558,14556}
Catalog No. NC1591300