Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC MSC Jumbo Envelope, 14 x 18 in, 28 lb., 250/Pk.

Supplier:  MSC 39577317

Encompass

This jumbo mailing envelope is designed to protect oversized printed items during transit or storage. Constructed from durable 28 lb. paper stock, its generous 14 x 18 inch dimensions accommodate large documents, and ungummed flaps allow for convenient reuse.

  • Jumbo envelope type for large items
  • White color for professional appearance
  • Made from durable 28 lb. paper stock
  • Ungummed flaps for reusability and flat storage
  • Measures 14 in wide and 18 in long

Catalog No. 50-191-8145


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.