Learn More
Hach Company Universal junction box for 4-20 mA - for AS950 samplers
Supplier:  Hach Company 9501000
Encompass
The Universal Junction Box for 4-20 mA is an accessory used to connect 0/4-20 mA transmitters or flow meters to a sampler auxiliary input. It includes an auxiliary cable and is documented as an accessory for AS950 series samplers.
  • Provides connection for 0/4-20 mA transmitters and flow meters.
  • Includes an auxiliary cable for direct connection to the sampler.
  • Allows integration with a sampler auxiliary I/O for flow pacing or analog inputs.
  • Supports simple field wiring to the sampler auxiliary port.
Catalog No. NC2110949